Learn More
Teknova HBS-EP Buffer (10mM Hepes, 150mM NaCl, 3mM EDTA, 0.005% Tween-20) 200mL, Sterile. 10 pack.
SDP
Catalog No. 502387396
Encompass
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.
Catalog No. 50-238-7396 Supplier Teknova Supplier No. H802010PK
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.