Learn More
ATHENA ES HBS w/Phenol Red, 500 ml
Catalog No. 5000166062
Encompass_Preferred
Hepes Buffered Saline is available with or without Phenol Red. It is an isotonic saline buffered with Hepes, for use in rinsing cell monolayers prior to cell dissociation using PETTM or washing harvested cells.
Catalog No. 50-001-66062 Supplier ATHENA ES Supplier No. 0408
Hepes Buffered Saline is available with or without Phenol Red. It is an isotonic saline buffered with Hepes, for use in rinsing cell monolayers prior to cell dissociation using PETTM or washing harvested cells.