Learn More
Aviva Systems Biology Corp HBS1L Antibody - C-terminal region (ARP52212_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52212_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HBS1L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HBS1L. Alternate names: DKFZp686L13262, EF-1a, ERFS, HBS1, HSPC276ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60701