Learn More
Cayman Chemical Gossypin, Purity: ≥95%, CAS: 652-78-8, 100 mg
SDP
Catalog No. 502950400
Encompass_Preferred
A flavonoid glycoside with diverse biological activities; inhibits RANKL-induced osteoclastogenesis in RAW 264.7 cells at 5 µM; inhibits Aurora B kinase (IC50 = 11.07 µM in a cell-free assay), as well as Aurora A kinase and RSK2 at 20 µM; induces cell cycle arrest at the G2/M phase and apoptosis in HGC-27 cells; decreases LDH release induced by D,L-buthionine (S,R)-sulfoximine in primary rat cortical cells (IC50 = 7.4 µg/ml); reduces acetic acid-induced writhing in mice
Catalog No. 50-295-0400 Supplier Cayman Chemical Supplier No. 34142100 MG
A flavonoid glycoside with diverse biological activities; inhibits RANKL-induced osteoclastogenesis in RAW 264.7 cells at 5 µM; inhibits Aurora B kinase (IC50 = 11.07 µM in a cell-free assay), as well as Aurora A kinase and RSK2 at 20 µM; induces cell cycle arrest at the G2/M phase and apoptosis in HGC-27 cells; decreases LDH release induced by D,L-buthionine (S,R)-sulfoximine in primary rat cortical cells (IC50 = 7.4 µg/ml); reduces acetic acid-induced writhing in mice