Learn More
Promocell GmbH PromoCell HBSMC-c 500,000 Human Bronchial Smooth Muscle Cells, cryopreserved
Catalog No. 50306033
Encompass
Primary Human Bronchial Smooth Muscle Cells (HBSMC) are isolated from the bronchi from single donors and stain positive for smooth muscle α-actin. They connect the endings of the bronchial cartilage and consequently regulate the airway lumen. Under pathological conditions like asthma, the amount of smooth muscle cells in the bronchi walls is increased leading to smooth muscle hypertrophy or hyperplasia. Hence, HBSMC are suitable for in vitro studies of asthma and other pulmonary diseases.Our HBSMC are now also available from HLA-typed and COPD/Asthma donors.
Catalog No. 50-306-033 Supplier Promocell GmbH Supplier No. C12561
Primary Human Bronchial Smooth Muscle Cells (HBSMC) are isolated from the bronchi from single donors and stain positive for smooth muscle α-actin. They connect the endings of the bronchial cartilage and consequently regulate the airway lumen. Under pathological conditions like asthma, the amount of smooth muscle cells in the bronchi walls is increased leading to smooth muscle hypertrophy or hyperplasia. Hence, HBSMC are suitable for in vitro studies of asthma and other pulmonary diseases.Our HBSMC are now also available from HLA-typed and COPD/Asthma donors.