Learn More
FUJIFILM BIOSCIENCES INC HBSS 10X-Hanks balan Salt sol
Catalog No. 5000450261
Encompass
HBSS 10X Hanks Balanced Salt Solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions
Catalog No. 50-004-50261 Supplier FUJIFILM BIOSCIENCES INC Supplier No. 9230500ML
Add to Cart
Add to Cart
HBSS 10X Hanks Balanced Salt Solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions