Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc HBSS (10X) without Sodium Bicarbonate, 500 mL
SDP

Catalog No. 507517572
Encompass_Preferred

HBSS provides a buffering system to maintain the physiological pH range and osmotic balance of the culture media. It provides cells with a source of water and essential inorganic ions, and carbon.

With Calcium and Magnesium
With Phenol Red
With Sodium Bicarbonate

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7572 Supplier Quality Biological Inc Supplier No. 119065101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HBSS provides a buffering system to maintain the physiological pH range and osmotic balance of the culture media. It provides cells with a source of water and essential inorganic ions, and carbon.

With Calcium and Magnesium
With Phenol Red
With Sodium Bicarbonate

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.