Learn More
FUJIFILM Biosciences HBSS 1X-Hanks balan Salt sol
Catalog No. 5000450251
Encompass
HBSS 1X Hanks Balanced Salt Solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions
Catalog No. 50-004-50251 Supplier FUJIFILM Biosciences Supplier No. 9220500ML
HBSS 1X Hanks Balanced Salt Solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions