Learn More
FUJIFILM BIOSCIENCES INC HBSS 1X-Hanks Balancd Salt Sol
Supplier:  FUJIFILM BIOSCIENCES INC 9228500ML
Encompass
HBSS 1X Hanks Balanced Salt Solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions
Catalog No. 50-004-50195