Learn More
FUJIFILM Biosciences HBSS 1X-Hanks Balancd Salt Sol
Catalog No. 5000450298
Encompass
HBSS 1X Hanks Balanced Salt Solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions
Catalog No. 50-004-50298 Supplier FUJIFILM Biosciences Supplier No. 9228100ML
HBSS 1X Hanks Balanced Salt Solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions