Learn More
FUJIFILM Biosciences HBSS 1X-Hanks Balanced Salt So
Catalog No. 5000450250
Encompass
HBSS 1X Hanks Balanced Salt Solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions
Catalog No. 50-004-50250 Supplier FUJIFILM Biosciences Supplier No. 9220100ML
HBSS 1X Hanks Balanced Salt Solution is designed to maintain pH and osmotic balance and to provide cells with water and essential inorganic ions