Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc HBSS without Calcium, Magnesium and Phenol Red, 500 mL
SDP

Catalog No. 50983209
Encompass_Preferred

Hanks Balanced Salt Solution (HBSS) provides a buffering system to maintain the physiological pH range and osmotic balance of the culture media. It provides cells with a source of water, essential inorganic ions, and carbohydrate as an energy source. Useful for diluting cells for counting. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

Without Calcium
Without Magnesium
Without Phenol Red

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-983-209 Supplier Quality Biological Inc Supplier No. 114062101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hanks Balanced Salt Solution (HBSS) provides a buffering system to maintain the physiological pH range and osmotic balance of the culture media. It provides cells with a source of water, essential inorganic ions, and carbohydrate as an energy source. Useful for diluting cells for counting. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

Without Calcium
Without Magnesium
Without Phenol Red

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.