Learn More
Quality Biological Inc HBSS without Phenol Red, 500 mL
SDP
Catalog No. 507517384
Encompass_Preferred
Hanks¿ Balanced Salt Solution (HBSS) provides a buffering system to maintain the physiological pH range and osmotic balance of the culture media. ¿ Provides cells with a source of water, essential inorganic ions, and carbohydrate as an energy source. ¿ Useful for diluting cells for counting. ¿ Formulations with calcium and magnesium support cell adhesion and are generally used as transport media, or for reagent preparation. ¿ Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 24 months
Catalog No. 50-751-7384 Supplier Quality Biological Inc Supplier No. 114061101
Hanks¿ Balanced Salt Solution (HBSS) provides a buffering system to maintain the physiological pH range and osmotic balance of the culture media. ¿ Provides cells with a source of water, essential inorganic ions, and carbohydrate as an energy source. ¿ Useful for diluting cells for counting. ¿ Formulations with calcium and magnesium support cell adhesion and are generally used as transport media, or for reagent preparation. ¿ Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 24 months