Learn More
STEMCELL Technologies HBSS, Modified (Without Ca++ and Mg++), Size: 500 mL
SDP
Catalog No. NC1100320
Encompass
Hanks’ Balanced Salt Solution (HBSS) can be used for diluting, washing, irrigating, or as a transporting solution for cell or tissue culture. It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media, and provides cells with a source of water and essential inorganic ions, and a carbohydrate as an energy source. Selection of suitable nutrient medium is dependent on type of cell, culture conditions, and degree of chemical definition required for the cell culture application.
Catalog No. NC1100320 Supplier STEMCELL Technologies Supplier No. 37250
Hanks’ Balanced Salt Solution (HBSS) can be used for diluting, washing, irrigating, or as a transporting solution for cell or tissue culture. It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media, and provides cells with a source of water and essential inorganic ions, and a carbohydrate as an energy source. Selection of suitable nutrient medium is dependent on type of cell, culture conditions, and degree of chemical definition required for the cell culture application.