Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ HBSS, calcium, magnesium, no phenol red
GREENER_CHOICE

Catalog No. 14025076
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1000 mL
10 x 500 mL
6 x 1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-025-076 1000 mL
14-025-092 500 mL
14-025-134 10 x 500 mL
14-025-126 6 x 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 14-025-076 Supplier Gibco™ Supplier No. 14025076
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media, or for reagent preparation.

We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.

This HBSS is modified as follows:

With: Calcium, Magnesium, Glucose

Without: Phenol Red, Sodium Pyruvate

The complete formulation is available.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hanks' Balanced Salt Solution (HBSS)
Concentration 1 X
For Use With (Application) Mammalian Cell Culture
pH 6.7 - 7.8
Physical Form Liquid
Product Line Gibco
Quantity 1000 mL
Solution Type Hank's Balanced Salt
Sterility Sterile-filtered
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Osmolality 270 - 305 mOsm/kg
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.