Learn More
Sino Biological Hepatitis B Virus (HBV)(ayw/France/Tiollais/1979) Capsid protein (His Tag)
Supplier:  Sino Biological 40573V08E100
Encompass_Preferred
A DNA sequence encoding the Hepatitis B Virus (HBV)(ayw/France/Tiollais/1979) Capsid protein (P03146-1) (Met1-Val149) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3698