Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HBV Nucleic Acid Purification Kit (Pre-filled)

Catalog No. 89339140 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-140 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-140 Supplier Abnova™ Supplier No. KA5518
Add to Cart
Add to Cart

HBV Nucleic Acid Purification Kit (Pre-filled) is designed specifically for the optimal automated extraction of nuclei acid from HBV on the SpiralPipet™ instrument.

Specifications

Content And Storage Store at room temperature.
Format Kit
Isolation Technology Magnetic Bead
Product Type Nucleic Acid Purification Kit
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.