Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix HBV Seroconversion Panels - 20 to 40 Member Panel
SDP

Catalog No. 22157172 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
23 x 1 mL
31 x 1 mL
35 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-172 31 x 1 mL
22-156-976 23 x 1 mL
22-156-980 35 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-157-172 Supplier ZeptoMetrix Supplier No. HBV9093
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Donor Demographics: Donor source plasma was collected from a U.S. licensed blood collection facility. Donor was un-treated and asymptomatic throughout the collection period.

Specifications

Certifications/Compliance FDA
Content And Storage -20°C
Product Type Seroconversion Reference Panel
Quantity 31 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.