Learn More
iXCells Biotechnologies Human Brain Vascular Pericytes (HBVP)
Supplier:  iXCells Biotechnologies 10HU031
Encompass_Preferred
iXCells Biotechnologies provides high quality Human Brain Vascular Pericytes (HBVP), which are isolated from human brain and cryopreserved at P2, with >0.5 million cells in each vial. These HBVP are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi. They can further expand in Human Pericyte Growth Medium under the condition suggested by iXCells Biotechnologies.
Catalog No. 50-107-8048