Learn More
Medchemexpress LLC HY-17540 5mg Medchemexpress, HBX 19818 CAS:1426944-49-1 Purity:>98%
SDP
Catalog No. 501932804
Encompass_Preferred
Medchemexpress, HY-17540 5mg HBX 19818 CAS:1426944-49-1 HBX 19818 is a specific inhibitor of ubiquitin-specific protease 7 (USP7) , with an IC 50 of 28.1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2804 Supplier Medchemexpress LLC Supplier No. HY175405MG
Medchemexpress, HY-17540 5mg HBX 19818 CAS:1426944-49-1 HBX 19818 is a specific inhibitor of ubiquitin-specific protease 7 (USP7) , with an IC 50 of 28.1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.