Learn More
Enzo Life Sciences Hepatitis Bx protein monoclonal antibody (X36C) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804278C100
Encompass_Preferred
Clone: X36C. Isotype: IgG1. Immunogen: Recombinant HBx protein (hepatitis Bx protein). Species reactivity: Hepatitis B virus. Specificity: Recognizes HBx. Detects a band of ~17kDa by Western blot. Applications: ELISA, IP, WB. Formulation: Liquid. In PBS containing 0.05% sodium azide.
Catalog No. 50-200-9853