Learn More
Aviva Systems Biology Corp HBXIP Antibody - N-terminal region (ARP52178_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52178_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HBXIP. It was validated on Western Blot. . Alternate names: MGC71071, XIP, HBXIPApplicationWB. Species reactivity: Human
Catalog No. 50-285-655