Learn More
Creative Biomart Recombinant Human LAMTOR5 Antigen, His-tagged
Supplier:  Creative Biomart HBV147
Encompass_Preferred
HBXIP Human Recombinant produced in E. coli is a single polypeptide chain containing 197 amino acids (aa 1-173) and having a molecular mass of 20.7 kDa.HBXIP is fused to a 24 amino acid His-tag at N-t
Catalog No. 50-208-4525
 Restricted Item