Learn More
Aviva Systems Biology Corp HBZ Antibody - N-terminal region (ARP42231_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42231_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HBZ. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HBZ. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58154