Learn More
Apexbio Technology LLC HC-030031, 10mg. Cas: 349085-38-7 MFCD: MFCD02007349. TRPA1 channel blocker,potent and selective
SDP
Supplier:  Apexbio Technology LLC B210010
Encompass_Preferred
HC-030031 is a selective antagonist of transient receptor potential ankyrin subfamily, member 1(TRPA1) with IC50 value of 4.9M [1].TRP channels are found to take participate in the response of chemical stimuli and in mechanoreception. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5307