Learn More
Enzo Life Sciences HC-030031 (10mg). CAS: 349085-38-7
SDP
Catalog No. 502010378
Encompass_Preferred
A potent and selective TRPA1 inhibitor. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0378 Supplier Enzo Life Sciences Supplier No. BMLCA2410010
A potent and selective TRPA1 inhibitor. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25 mg/ml). Long Term Storage: Ambient.