Learn More
Medchemexpress LLC HY-112302 5mg Medchemexpress, HC-070 CAS:1628291-95-1 Purity:>98%
SDP
Catalog No. 501969077
Encompass_Preferred
Medchemexpress, HY-112302 5mg HC-070 CAS:1628291-95-1 HC-070 is an antagonist of TRPC4/TRPC5, with IC50s of 9.3 nM and 46 nM for hTRPC5 and hTRPC4 in cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9077 Supplier Medchemexpress LLC Supplier No. HY1123025MG
Medchemexpress, HY-112302 5mg HC-070 CAS:1628291-95-1 HC-070 is an antagonist of TRPC4/TRPC5, with IC50s of 9.3 nM and 46 nM for hTRPC5 and hTRPC4 in cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.