Learn More
Apexbio Technology LLC HyperChrom Glutathione 4FF Agarose 10ml
SDP
Catalog No. 5000063139
Encompass_Preferred
HyperChrom Glutathione 4FF Agarose is an affinity chromatography medium designed for purifying glutathione S-transferase (GST)-tagged recombinant proteins The medium employs immobilized glutathione ligands coupled to agarose matrices selectively interacting through specific affinity binding with GST fusion proteins enabling purification under mild conditions Beyond GST-tagged proteins it can be utilized to isolate other proteins dependent on glutathione-mediated interactions or native glutathione S-transferases Its high binding capacity scalability and regenerability provide versatility for protein purification applications in biomedical research laboratories
Catalog No. 50-000-63139 Supplier Apexbio Technology LLC Supplier No. PC200310
HyperChrom Glutathione 4FF Agarose is an affinity chromatography medium designed for purifying glutathione S-transferase (GST)-tagged recombinant proteins The medium employs immobilized glutathione ligands coupled to agarose matrices selectively interacting through specific affinity binding with GST fusion proteins enabling purification under mild conditions Beyond GST-tagged proteins it can be utilized to isolate other proteins dependent on glutathione-mediated interactions or native glutathione S-transferases Its high binding capacity scalability and regenerability provide versatility for protein purification applications in biomedical research laboratories