Learn More
Apexbio Technology LLC HyperChrom Heparin FF Agarose 10ml
SDP
Catalog No. 5000062914
Encompass_Preferred
HyperChrom Heparin FF Agarose is an affinity chromatography resin composed of heparin covalently immobilized onto a cross-linked agarose support Heparin a naturally occurring glycosaminoglycan acts as an affinity ligand based on its specific interactions with various biomolecules including coagulation factors antithrombin III growth factors interferons lipoprotein lipases nucleic acid-binding proteins and steroid receptors This resin is employed extensively in biomedical research for the purification and isolation of these biological entities
Catalog No. 50-000-62914 Supplier Apexbio Technology LLC Supplier No. PC200810
HyperChrom Heparin FF Agarose is an affinity chromatography resin composed of heparin covalently immobilized onto a cross-linked agarose support Heparin a naturally occurring glycosaminoglycan acts as an affinity ligand based on its specific interactions with various biomolecules including coagulation factors antithrombin III growth factors interferons lipoprotein lipases nucleic acid-binding proteins and steroid receptors This resin is employed extensively in biomedical research for the purification and isolation of these biological entities