Learn More
Apexbio Technology LLC HyperChrom Ni-NTA FF Agarose 10ml
SDP
Catalog No. 5000062912
Encompass_Preferred
HyperChrom Ni-NTA FF Agarose is an affinity chromatography resin composed of highly cross-linked agarose matrix chemically functionalized with nitrilotriacetic acid (NTA) ligands that chelate nickel ions (Ni ) These chelated nickel ions exhibit a stable octahedral coordination state enabling selective binding to proteins containing histidine residues particularly polyhistidine-tagged recombinant proteins The resin demonstrates resistance to reducing agents denaturants and elevated pressures (up to 0 3 MPa) facilitating protein purification under stringent conditions and at increased flow rates Due to its high binding capacity stability ease of regeneration and cost-effectiveness it is routinely used in research and industrial-scale purification processes targeting His-tagged proteins
Catalog No. 50-000-62912 Supplier Apexbio Technology LLC Supplier No. PC200110
HyperChrom Ni-NTA FF Agarose is an affinity chromatography resin composed of highly cross-linked agarose matrix chemically functionalized with nitrilotriacetic acid (NTA) ligands that chelate nickel ions (Ni ) These chelated nickel ions exhibit a stable octahedral coordination state enabling selective binding to proteins containing histidine residues particularly polyhistidine-tagged recombinant proteins The resin demonstrates resistance to reducing agents denaturants and elevated pressures (up to 0 3 MPa) facilitating protein purification under stringent conditions and at increased flow rates Due to its high binding capacity stability ease of regeneration and cost-effectiveness it is routinely used in research and industrial-scale purification processes targeting His-tagged proteins