Learn More
Apexbio Technology LLC HyperChrom Phenyl HP Agarose 100ml
SDP
Catalog No. 5000062678
Encompass_Preferred
HyperChrom Phenyl HP Agarose is a hydrophobic interaction chromatography (HIC) resin designed for separation and purification of biomolecules based on differences in their hydrophobic properties The functional medium utilizes phenyl ligands immobilized onto a highly cross-linked agarose support enabling biomolecule interaction under defined conditions Its chemically modified hydrophilic agarose backbone provides mechanical robustness and supports high-flow chromatography processes HyperChrom Phenyl HP Agarose is mainly applied in purification of recombinant proteins antibodies and viral vaccines particularly suited for bioseparation and downstream biopharmaceutical applications
Catalog No. 50-000-62678 Supplier Apexbio Technology LLC Supplier No. PC2019100
HyperChrom Phenyl HP Agarose is a hydrophobic interaction chromatography (HIC) resin designed for separation and purification of biomolecules based on differences in their hydrophobic properties The functional medium utilizes phenyl ligands immobilized onto a highly cross-linked agarose support enabling biomolecule interaction under defined conditions Its chemically modified hydrophilic agarose backbone provides mechanical robustness and supports high-flow chromatography processes HyperChrom Phenyl HP Agarose is mainly applied in purification of recombinant proteins antibodies and viral vaccines particularly suited for bioseparation and downstream biopharmaceutical applications