Learn More
Apexbio Technology LLC HyperChrom Protein G 4FF Agarose 10ml
SDP
Catalog No. 5000064360
Encompass_Preferred
HyperChrom Protein G 4FF Agarose is an affinity chromatography resin produced by covalently coupling Protein G onto a Chromstar matrix via cyanogen bromide (CNBr)-mediated activation Protein G selectively binds the Fc domain of various mammalian IgG antibodies and interacts weakly with Fab regions offering broader specificity compared to Protein A This resin is utilized primarily in biomedical research to isolate and purify immunoglobulins antibody fragments and immune complexes from complex biological matrices including serum and cell culture supernatants
Catalog No. 50-000-64360 Supplier Apexbio Technology LLC Supplier No. PC200610
HyperChrom Protein G 4FF Agarose is an affinity chromatography resin produced by covalently coupling Protein G onto a Chromstar matrix via cyanogen bromide (CNBr)-mediated activation Protein G selectively binds the Fc domain of various mammalian IgG antibodies and interacts weakly with Fab regions offering broader specificity compared to Protein A This resin is utilized primarily in biomedical research to isolate and purify immunoglobulins antibody fragments and immune complexes from complex biological matrices including serum and cell culture supernatants