Learn More
Ubpbio HC Streptavidin Agarose, 5ML
Supplier:  Ubpbio P30705
Encompass_Preferred
High Capacity Streptavidin Agarose Resin is an affinity chromatography medium designed for easy, one-step purification of biotinylated peptides, antibodies, lectins, etc from samples. The purified recombinant streptavidin is covalently coupling to 4% highly cross-linked agarose. The coupling is optimized to give high binding capacity for biotinylated molecules. The total binding capacity of Streptavidin Resin is more than 120 nmol of D-Biotin/ml settled resin.
Catalog No. 50-196-5141