Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HC-Toxin | 83209-65-8 | 99.3% | 500 UG
SDP

Catalog No. 5000341582
Encompass_Preferred

HC-Toxin is a cyclic tetrapeptide acting as a potent HDAC inhibitor. It is used in laboratory research to induce tumor cell apoptosis and exhibits anticancer effects.

  • Potent HDAC inhibitor with an IC50 of 30 nM
  • Induces tumor cell apoptosis
  • Exhibits anticancer effects
  • Stable under recommended storage conditions
  • Appearance: Off-white to light yellow solid
  • Molecular weight: 436.50
  • Molecular formula: C21H32N4O6
  • Solubility of ≥ 10 mg/mL in DMSO
  • Suitable for laboratory chemical applications
  • Store at -20°C, protected from light

Catalog No. 50-003-41582 Supplier Medchemexpress LLC Supplier No. HY126856500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HC-Toxin is a cyclic tetrapeptide acting as a potent HDAC inhibitor. It is used in laboratory research to induce tumor cell apoptosis and exhibits anticancer effects.

  • Potent HDAC inhibitor with an IC50 of 30 nM
  • Induces tumor cell apoptosis
  • Exhibits anticancer effects
  • Stable under recommended storage conditions
  • Appearance: Off-white to light yellow solid
  • Molecular weight: 436.50
  • Molecular formula: C21H32N4O6
  • Solubility of ≥ 10 mg/mL in DMSO
  • Suitable for laboratory chemical applications
  • Store at -20°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.