Learn More
Cayman Chemical HC Toxin, Purity: ≥95%, CAS: 83209-65-8, 5 mg
SDP
Catalog No. 502890252
Encompass_Preferred
A cell-permeable, reversible inhibitor of HDACs (IC50 = 30 nM)
Catalog No. 50-289-0252 Supplier Cayman Chemical Supplier No. 105765 MG
A cell-permeable, reversible inhibitor of HDACs (IC50 = 30 nM)