Learn More
Cayman Chemical HC Toxin Inhibitors
SDP
Catalog No. 502890515
Encompass_Preferred
A cell-permeable, reversible inhibitor of HDACs (IC50 = 30 nM)
Catalog No. 50-289-0515 Supplier Cayman Chemical Supplier No. 10576500 UG
Add to Cart
Add to Cart
A cell-permeable, reversible inhibitor of HDACs (IC50 = 30 nM)