Learn More
Cayman Chemical HC Toxin, Purity: ≥95%, CAS: 83209-65-8, 1 mg
SDP
Catalog No. 502888919
Encompass_Preferred
A cell-permeable, reversible inhibitor of HDACs (IC50 = 30 nM)
Catalog No. 50-288-8919 Supplier Cayman Chemical Supplier No. 105761 MG
A cell-permeable, reversible inhibitor of HDACs (IC50 = 30 nM)