Learn More
KELLEY SUPPLY INC HC10 Alkaline Cleaner | 50lb
Catalog No. NC1792913
Encompass
Ecolab HC-10 Chlorinated Kleer-Mor is an alkaline detergent for manual cleaning of processing and dairy equipment, inculding: bulk raw and pasteurized tanks, transfer lines, pumps and associated equipment. This stable, organic, and chlorine-releasing compound quickly attacks and removes protein soils making manual cleaning easier and faster.
Catalog No. NC1792913 Supplier KELLEY SUPPLY INC Supplier No. C310080PP
Ecolab HC-10 Chlorinated Kleer-Mor is an alkaline detergent for manual cleaning of processing and dairy equipment, inculding: bulk raw and pasteurized tanks, transfer lines, pumps and associated equipment. This stable, organic, and chlorine-releasing compound quickly attacks and removes protein soils making manual cleaning easier and faster.