Learn More
Mitegen HC12 Liquid Nitrogen Refrigerator By IC Biomedical For Sample Storage in Liquid Nitrogen, 12 Liter Capacity
SDP
Catalog No. 5000494999
Encompass
The IC Biomedical HC12 Liquid Nitrogen Refrigerator is a 12-liter cryogenic freezer designed for storing a variety of materials at extremely low temperatures. This unit provides temperatures of -196°C at the liquid surface and -190°C at the canister top. It has a large storage capacity with superior vacuum performance and super insulation for maximum holding times. The refrigerator features a rugged, high-strength aluminum body, a convenient canister numbered index location ring, and a tapered internal spider. A roller base TW-R018 is available for improved mobility. The unit has a static holding time of 60 days, an evaporation rate of 0.20 liters per day, and a 12-liter liquid nitrogen capacity. The refrigerator comes with six thin-gauge stainless steel canisters. The manufacturer is ICBiomedical, and it comes with a 5-year vacuum warranty.
Catalog No. 50-004-94999 Supplier Mitegen Supplier No. TWHC12
The IC Biomedical HC12 Liquid Nitrogen Refrigerator is a 12-liter cryogenic freezer designed for storing a variety of materials at extremely low temperatures. This unit provides temperatures of -196°C at the liquid surface and -190°C at the canister top. It has a large storage capacity with superior vacuum performance and super insulation for maximum holding times. The refrigerator features a rugged, high-strength aluminum body, a convenient canister numbered index location ring, and a tapered internal spider. A roller base TW-R018 is available for improved mobility. The unit has a static holding time of 60 days, an evaporation rate of 0.20 liters per day, and a 12-liter liquid nitrogen capacity. The refrigerator comes with six thin-gauge stainless steel canisters. The manufacturer is ICBiomedical, and it comes with a 5-year vacuum warranty.