Learn More
Mitegen HC20 Liquid Nitrogen Refrigerator By IC Biomedical For Sample Storage in Liquid Nitrogen, 20 Liter Capacity
SDP
Catalog No. 5000494993
Encompass
The IC Biomedical HC20 Liquid Nitrogen Refrigerator is a 20-liter cryogenic freezer designed for storing various materials at cryogenic temperatures. The unit provides temperatures of -196ºC at the liquid surface and -190ºC at the canister top. This refrigerator features a large storage capacity with superior vacuum performance and super insulation, which provides maximum holding times. It has a new rugged construction, including a ribbed high-strength aluminum body, a magniformed necktube design, and a more durable paint finish. The refrigerator also has a convenient canister numbered index location ring and a tapered internal spider. The HC20 has a static holding time of 87 days and an evaporation rate of 0.23 liters per day. For added mobility, an optional roller base TW-R018-8C00 is available. The unit comes with six thin-gauge stainless steel canisters. The manufacturer is ICBiomedical, and it comes with a 5-year vacuum warranty.
Catalog No. 50-004-94993 Supplier Mitegen Supplier No. TWHC20
The IC Biomedical HC20 Liquid Nitrogen Refrigerator is a 20-liter cryogenic freezer designed for storing various materials at cryogenic temperatures. The unit provides temperatures of -196ºC at the liquid surface and -190ºC at the canister top. This refrigerator features a large storage capacity with superior vacuum performance and super insulation, which provides maximum holding times. It has a new rugged construction, including a ribbed high-strength aluminum body, a magniformed necktube design, and a more durable paint finish. The refrigerator also has a convenient canister numbered index location ring and a tapered internal spider. The HC20 has a static holding time of 87 days and an evaporation rate of 0.23 liters per day. For added mobility, an optional roller base TW-R018-8C00 is available. The unit comes with six thin-gauge stainless steel canisters. The manufacturer is ICBiomedical, and it comes with a 5-year vacuum warranty.