Learn More
Mitegen HC38 High Capacity Liquid Nitrogen Refrigerator By IC Biomedical For Sample Storage in Liquid Nitrogen, 38 Liter Capacity
SDP
Catalog No. 5000495000
Encompass
The IC Biomedical HC38 High Capacity Liquid Nitrogen Refrigerator is designed for storing a variety of materials at cryogenic temperatures, providing -196ºC at the liquid surface and -190ºC at the canister top. It features a large storage capacity with a 38-liter liquid nitrogen capacity. The refrigerator has a rugged construction, including a ribbed high-strength aluminum body, a magneformed necktube design, and a durable paint finish. Its superior vacuum performance with super insulation provides maximum holding times, with a static holding time of 120 days and an evaporation rate of 0.31 liters per day. For organization and ease of use, it includes a numbered index ring and a tapered internal canister locator. Security and mobility options include a lockable lid and an available roller base TW-R033-8C00 and optional low-liquid-level alarm TW-R038-8C35. The unit comes with 10 canisters, each measuring 2.75 x 11.0 in. The manufacturer is ICBiomedical.
Catalog No. 50-004-95000 Supplier Mitegen Supplier No. TWHC38
The IC Biomedical HC38 High Capacity Liquid Nitrogen Refrigerator is designed for storing a variety of materials at cryogenic temperatures, providing -196ºC at the liquid surface and -190ºC at the canister top. It features a large storage capacity with a 38-liter liquid nitrogen capacity. The refrigerator has a rugged construction, including a ribbed high-strength aluminum body, a magneformed necktube design, and a durable paint finish. Its superior vacuum performance with super insulation provides maximum holding times, with a static holding time of 120 days and an evaporation rate of 0.31 liters per day. For organization and ease of use, it includes a numbered index ring and a tapered internal canister locator. Security and mobility options include a lockable lid and an available roller base TW-R033-8C00 and optional low-liquid-level alarm TW-R038-8C35. The unit comes with 10 canisters, each measuring 2.75 x 11.0 in. The manufacturer is ICBiomedical.