Learn More
LifeSensors High Capacity Control Magnetic Beads (negative control)
SDP
Supplier:  LifeSensors UM0500M1000
Encompass_Preferred
For use in “pre-clearing” cell lysates, as well as a negative control for the pulldown of polyubiquitinated proteins in conjunction with magnetic beads-conjugated TUBEs. Control magnetic beads are generated by blocking an activated matrix (identical to that used for TUBEs conjugation) with ethanolamine. Control magnetic beads (1 mL) are supplied as a slurry in PBS, pH 7.2.
Catalog No. NC2285791