Learn More
Promocell GmbH PromoCell HCASMC-c 500,000 Human Coronary Artery Smooth Muscle Cells, cryopreserved
Supplier:  Promocell GmbH C12511
Encompass
Primary Human Coronary Artery Smooth Muscle Cells (HCASMC) are isolated from human coronary arteries and stain positive for smooth muscle α-actin. Coronary arteries supply the heart with blood and obstruction in atherosclerosis causes myocardial infarction.HCASMC are qualified to study the function of smooth muscle cells in the development of atherosclerotic plaques or in other diseases like hypertension. They are also useful for stent-graft compatibility testing.
Catalog No. 50-306-025