Learn More
Adam Equipment HCB 2202 - Highland Portable Precision Balances - Capacity: 2200g, Readability: 0.01g
Supplier:  Adam Equipment HCB 2202
Encompass_Preferred
The most complete and versatile precision balance for a wide range of applications. Adam Equipment’s Highland balances have what it takes for lab work, field use, and many industrial applications. They feature Adam’s unique patented ShockProtect overload protection to withstand up to 200kg, and Adam’s HandiCal internal calibration with built in mass. Calibrate whenever you want without external masses, or use your own certified masses for traceability. The smart housing design lets you safely stack Highland balances on top of one another for convenient storage and transportation. Both USB and RS232 interfaces are included with cables, and a rechargeable battery (adapter/charger included), removable draft shield, and brilliant backlit display with capacity tracking make Highland the best portable precision balance available today. Measures 6.7 inches wide x 9.6 inches deep x 3.1 inch high. Pan size is 4.7 inches diameter. Measures 6.7 inches wide x 9.6 deep x 3.1 tal
Catalog No. NC3964756