Learn More
Adam Equipment HCB 3001aM - Highland Approved Portable Precision Balances - Capacity: 3000g, Readability: d=0.5g e=0.5g
Catalog No. NC3964768
Encompass_Preferred
Elevate your precision weighing tasks with the Highland Approved Portable Precision Balances, designed to meet Class II requirements for retailers, jewelers, chemists, and pharmacists. These balances are NTEP-approved, ensuring compliance with U.S. legal and regulatory standards for pricing by weight.
Catalog No. NC3964768 Supplier Adam Equipment Supplier No. HCB 3001AM
Elevate your precision weighing tasks with the Highland Approved Portable Precision Balances, designed to meet Class II requirements for retailers, jewelers, chemists, and pharmacists. These balances are NTEP-approved, ensuring compliance with U.S. legal and regulatory standards for pricing by weight.