Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HCC-1/CCL14 Human 100ug
SDP

Catalog No. 5000195555
Encompass_Preferred

HCC-1 (CCL14) is a human CC chemokine involved in chemotaxis of monocytes, eosinophils, and T lymphocytes. This recombinant protein comprises the mature region T22-N93 (74 amino acids), is expressed in Escherichia coli, and is provided tag free for research applications such as chemotaxis and receptor signaling studies.

  • Recombinant human chemokine corresponding to mature region T22-N93 (74 aa).
  • Expressed in Escherichia coli.
  • Tag free, with no affinity tags present.
  • Suitable for in vitro chemotaxis and immune cell signaling assays.
  • Useful for receptor binding and cell migration studies.

Catalog No. 50-001-95555 Supplier Medchemexpress LLC Supplier No. HYP7195100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HCC-1 (CCL14) is a human CC chemokine involved in chemotaxis of monocytes, eosinophils, and T lymphocytes. This recombinant protein comprises the mature region T22-N93 (74 amino acids), is expressed in Escherichia coli, and is provided tag free for research applications such as chemotaxis and receptor signaling studies.

  • Recombinant human chemokine corresponding to mature region T22-N93 (74 aa).
  • Expressed in Escherichia coli.
  • Tag free, with no affinity tags present.
  • Suitable for in vitro chemotaxis and immune cell signaling assays.
  • Useful for receptor binding and cell migration studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.