Learn More
Medchemexpress LLC HY-P7195 10ug Medchemexpress, HCC-1/CCL14, Human CAS: Purity:>98%
SDP
Catalog No. 502083124
Encompass_Preferred
Medchemexpress, HY-P7195 10ug HCC-1/CCL14, Human CAS: Recombinant Human Hemofiltrate CC Chemokine-1/CCL14 has weak activities on human monocytes and acts via receptors that also recognize MIP-1 alpha. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3124 Supplier Medchemexpress LLC Supplier No. HYP719510UG
Medchemexpress, HY-P7195 10ug HCC-1/CCL14, Human CAS: Recombinant Human Hemofiltrate CC Chemokine-1/CCL14 has weak activities on human monocytes and acts via receptors that also recognize MIP-1 alpha. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.