Learn More
Medchemexpress LLC HY-P7197 10ug Medchemexpress, HCC-4/CCL16, Human CAS: Purity:>98%
SDP
Catalog No. 502083126
Encompass_Preferred
Medchemexpress, HY-P7197 10ug HCC-4/CCL16, Human CAS: Recombinant Human Hemofiltrate CC Chemokine-4/CCL16 specifically attracts lymphocytes, dendritic cells, and monocytes; increases their adhesive properties and has myelosuppressive activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3126 Supplier Medchemexpress LLC Supplier No. HYP719710UG
Medchemexpress, HY-P7197 10ug HCC-4/CCL16, Human CAS: Recombinant Human Hemofiltrate CC Chemokine-4/CCL16 specifically attracts lymphocytes, dendritic cells, and monocytes; increases their adhesive properties and has myelosuppressive activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.