Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HCC-4/CCL16 Human 2ug
SDP

Catalog No. 5000195959
Encompass_Preferred

Recombinant human HCC-4/CCL16 (Q24-Q120) is a tag-free chemokine produced in Escherichia coli and supplied lyophilized for research applications. It promotes lymphocyte, dendritic cell, and monocyte chemotaxis and adhesion, and is used to investigate inflammatory and cancer-related responses. The material is >95% pure by reducing SDS-PAGE and has a predicted molecular mass of 11.2 kDa.

  • Recombinant human chemokine (Q24-Q120) expressed in E. coli.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Tag free, lyophilized formulation from 20 mM PB, 150 mM NaCl, pH 7.4.
  • Predicted molecular mass 11.2 kDa (≈14 kDa by reducing SDS-PAGE).
  • Not recommended to reconstitute below 100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • Suitable for research on leukocyte chemotaxis, adhesion, and inflammatory pathways.

Catalog No. 50-001-95959 Supplier Medchemexpress LLC Supplier No. HYP71972UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human HCC-4/CCL16 (Q24-Q120) is a tag-free chemokine produced in Escherichia coli and supplied lyophilized for research applications. It promotes lymphocyte, dendritic cell, and monocyte chemotaxis and adhesion, and is used to investigate inflammatory and cancer-related responses. The material is >95% pure by reducing SDS-PAGE and has a predicted molecular mass of 11.2 kDa.

  • Recombinant human chemokine (Q24-Q120) expressed in E. coli.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Tag free, lyophilized formulation from 20 mM PB, 150 mM NaCl, pH 7.4.
  • Predicted molecular mass 11.2 kDa (≈14 kDa by reducing SDS-PAGE).
  • Not recommended to reconstitute below 100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • Suitable for research on leukocyte chemotaxis, adhesion, and inflammatory pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.