Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HCC-4/CCL16 Human 2ug
SDP

Supplier:  Medchemexpress LLC HYP71962UG

Encompass_Preferred

Recombinant human HCC-4/CCL16 is a purified chemokine expressed in CHO cells and supplied as a lyophilized powder for research applications; it has documented chemoattractant activity, low endotoxin, and recommended storage and handling to preserve activity.

  • Expressed in CHO cells for mammalian-like post-translational modifications.
  • Supplied as a lyophilized powder after extensive dialysis against PBS.
  • Low endotoxin level (<0.2 EU/μg) suitable for sensitive cell-based assays.
  • Demonstrated bioactivity with ED50 <1.5 μg/mL in a CCR1-based cell assay.
  • Approximate molecular weight 12 kDa as determined by SDS-PAGE.
  • Amino acid sequence and accession information provided for reproducibility.
  • Store at -20°C and freeze aliquots for extended stability.
  • Reconstitute at ≥100 μg/mL in ddH2O and consider adding carrier protein for long-term storage.

Catalog No. 50-001-96096


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.